Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetra-n-octylammonium bromide, 98+%
Click to view available options
Quantity:
10 g
50 g
250 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Tetra-n-octylammonium bromide is associated with benzethonium chloride and used as a disinfectant, in surface treatment and preservative in hospitals and industries. It is used as a surfactant involved in the synthesis of blue-luminescent silicon nanoparticles in order to get uniform particle size. It is utilized as a cationic additive in the preparation of membranes using cyclam derivative 1,4,8,11-tetra(n-octyl)-1,4,8,11-tetraazacyclotetradecane as an ionphore and dibutyl phthalate as plasticizers.
Solubility
Soluble in water, dichloromethane, chloroform, acetone and methanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Tetra-n-octylammonium bromide is associated with benzethonium chloride and used as a disinfectant, in surface treatment and preservative in hospitals and industries. It is used as a surfactant involved in the synthesis of blue-luminescent silicon nanoparticles in order to get uniform particle size. It is utilized as a cationic additive in the preparation of membranes using cyclam derivative 1,4,8,11-tetra(n-octyl)-1,4,8,11-tetraazacyclotetradecane as an ionphore and dibutyl phthalate as plasticizers.
Solubility
Soluble in water, dichloromethane, chloroform, acetone and methanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 94°C to 99°C |
| Assay Percent Range | 98% |
| Quantity | 10 g |
| Beilstein | 4169176 |
| Solubility Information | Soluble in water,dichloromethane,chloroform,acetone and methanol. |
| Formula Weight | 546.81 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Tetra-n-octylammonium Bromide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 238-936-9 |
| TSCA | No |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction