Alfa Aesar™ Tetra-n-propylammonium bromide, 98+%

Manufacturer: Alfa Aesar™ A115220E

Quantity 2500g

 View more versions of this product

Catalog No. AAA115220E

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1941-30-6
    Molecular Formula (CH3(CH2)2)4NBr
    Formula Weight 266.27
    MDL Number MFCD00011840
    Beilstein 3567846
    Chemical Name or Material Tetra-n-propylammonium bromide

    Specifications

    Formula Weight 266.27
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 1941-30-6
    Health Hazard 1 Warning
    TSCA TSCA
    MDL Number MFCD00011840
    Beilstein 3567846
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point ≈270°C (dec.)
    Quantity 2500g
    Molecular Formula (CH3(CH2)2)4NBr
    Chemical Name or Material Tetra-n-propylammonium bromide
    Assay ≥98%

    Description

    Applications
    Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.

    Solubility
    It is soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to Strong oxidizing agents. Stable under ordinary conditions (Hygroscopic)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit