Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraacetylethylenediamine, 90%

Catalog No. p-195577
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle

CAS: 10543-57-4 | C10H16N2O4 | 228.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10543-57-4
Molecular Formula C10H16N2O4
Molecular Weight (g/mol) 228.25
MDL Number MFCD00014967
InChI Key BGRWYDHXPHLNKA-UHFFFAOYSA-N
Synonym tetraacetylethylenediamine, n,n,n',n'-tetraacetylethylenediamine, taed, tetracetylethylenediamine, acetamide, n,n'-1,2-ethanediylbis n-acetyl, n,n'-ethylenebis n-acetylacetamide, unii-p411ed0n2b, n,n'-ethane-1,2-diyl bis n-acetylacetamide, n-acetyl-n-2-diacetylamino ethyl acetamide
PubChem CID 66347
IUPAC Name N-acetyl-N-[2-(diacetylamino)ethyl]acetamide
SMILES CC(=O)N(CCN(C(=O)C)C(=O)C)C(=O)C

Specifications

Melting Point 149°C to 154°C
Color White to Beige
Boiling Point 140°C (2.0 hPa)
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 88% min
Packaging Plastic bottle
Quantity 50 g
Merck Index 15, 9158
Solubility Information Solubility in water: ca 1g/L (20°C). Other solubilities: slightly soluble in acetone,moderately soluble in chloroform,methylenechloride
Formula Weight 228.25
Percent Purity 90%
Physical Form Granular Powder
Chemical Name or Material Tetraacetylethylenediamine, 90%
Show More Show Less
EINECSNumber 234-123-8
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.