Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraammineplatinum(II) chloride monohydrate, Premion™, 99.995% (metals basis)

Catalog No. p-7048545
Click to view available options
Quantity:
1 g
5 g

CAS: 13933-33-0 | Cl2H14N4OPt | 352.12 g/mol

Tetraammineplatinum(II) chloride is used as a catalyst in the chemical synthesis and a precursor for platinum catalysts. This complex has pharmaceutical applications and used in proteomics research. It is used in the preparation of Pt catalysts supported on reduced graphite oxide (RGO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13933-33-0
Molecular Formula Cl2H14N4OPt
Molecular Weight (g/mol) 352.12
MDL Number MFCD00149947
InChI Key GWBDNMCYHWRSOH-UHFFFAOYSA-L
Synonym azane;platinum 2+ ;dichloride;hydrate, tetrammineplatinum ii chloride hydrate, tetraammineplatinum ii chloride monohydrate, platinum 2+ ion tetraamine hydrate dichloride, platinum 2+ tetraamine hydrate dichloride, platinum 2+ chloride-ammonia-water 1/2/4/1, tetraamineplatinium ii chloride
PubChem CID 21946612
IUPAC Name azane;platinum(2+);dichloride;hydrate
SMILES N.N.N.N.O.[Cl-].[Cl-].[Pt++]

Specifications

Melting Point 250°C (dec.)
Color White through Cream
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water
Formula Weight 352.13 (334.11 Anhydrous)
Percent Purity 99.995%
Grade Premion™
Sensitivity Hygroscopic
Density 2.74g/mL
Assay (metals basis)
Chemical Name or Material Tetraammineplatinum(II) chloride monohydrate
Show More Show Less
Hazard Category H290-H315-H319
Hazard Statement GHS H Statement
H290-H315-H319
May be corrosive to metals.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
EINECSNumber 237-706-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.