Learn More
Tetraammineplatinum(II) nitrate solution, Pt 3-4% w/w (cont. Pt)
Description
Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 0.5 g |
| Linear Formula | (NH3)4Pt(NO3)2 |
| UN Number | UN3218 |
| Solubility Information | Miscible with water. |
| Formula Weight | 387.21 |
| Concentration or Composition (by Analyte or Components) | Pt 3 to 4% w/w (cont. Pt) |
| Density | 1.05g/mL |
| Chemical Name or Material | Tetraammineplatinum(II) nitrate solution |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H240-H272 Heating may cause an explosion. May intensify fire oxidizer. |
| Precautionary Statement | GHS P Statement P221-P210-P370+P378a-P410-P411a+P235-P501a Take any precaution to avoid mixing with combustibles. Keep away from heat/sparks/open flames/hot surfaces. No smoking. In case of fire: Use for extinction: CO2, powder or water spray. Protect from sunlight. Store at temperatures not exceeding 392°F. Keep cool. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S . |
| EINECSNumber | 243-929-9 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.