Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraammineplatinum(II) nitrate solution, Pt 3-4% w/w (cont. Pt)

Catalog No. AA12167NA
Click to view available options
Quantity:
0.5 g
2 g

CAS: 20634-12-2 | H12N6O6Pt | 387.22 g/mol

Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20634-12-2
Molecular Formula H12N6O6Pt
Molecular Weight (g/mol) 387.22
MDL Number MFCD00011624
InChI Key RBAKORNXYLGSJB-UHFFFAOYSA-N
Synonym tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt
PubChem CID 146543
IUPAC Name azane;platinum(2+);dinitrate
SMILES N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Colorless
Physical Form Liquid
Quantity 2 g
Linear Formula (NH3)4Pt(NO3)2
UN Number UN3218
Solubility Information Miscible with water.
Formula Weight 387.21
Concentration or Composition (by Analyte or Components) Pt 3 to 4% w/w (cont. Pt)
Density 1.05g/mL
Chemical Name or Material Tetraammineplatinum(II) nitrate solution
Hazard Category Danger
Hazard Statement GHS H Statement
H240-H272
Heating may cause an explosion.
May intensify fire
oxidizer.
Precautionary Statement GHS P Statement
P221-P210-P370+P378a-P410-P411a+P235-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
In case of fire: Use for extinction: CO2, powder or water spray.
Protect from sunlight.
Store at temperatures not exceeding 392°F.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S .
EINECSNumber 243-929-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.