Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutyl germanium, 98%, Thermo Scientific™

Catalog No. AC321030050
Click to view available options
:
5g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1067-42-1
Density 0.9300g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C16H36Ge
Linear Formula [CH3(CH2)3]4Ge
Quantity 5g
MDL Number MFCD00015224
Synonym tetrabutylgermanium, germane, tetrabutyl, tetra-n-butylgermane, tetra-n-butylgermanium, tetrabutyl-german, tetrabuthylgermanium, tetrabutyl germanium
InChI Key HDVLQIDIYKIVRE-UHFFFAOYSA-N
SMILES CCCC[Ge](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylgermane
Molecular Weight (g/mol) 301.094
PubChem CID 70599
Formula Weight 301.06
Percent Purity ≥97.5%
Chemical Name or Material Tetrabutyl germanium
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-929-3
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.