Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Bifluoride 95.0+%, TCI America™
SDP

Catalog No. T129525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T129525G 25 g
T12955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T129525G Supplier TCI America Supplier No. T129525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23868-34-0
Molecular Formula C16H37F2N
Molecular Weight (g/mol) 281.476
MDL Number MFCD00077877
InChI Key ZHBDKVWQJKYIFF-UHFFFAOYSA-M
Synonym Tetrabutylammonium Hydrogen Difluoride
PubChem CID 10891295
IUPAC Name tetrabutylazanium;fluoride;hydrofluoride
SMILES CCCC[N+](CCCC)(CCCC)CCCC.F.[F-]

Specifications

Quantity 25 g
UN Number 1760
Formula Weight 281.48
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Bifluoride
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.