Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
429-41-4,109-99-9,7732-18-5
C16H36FN
261.46
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylammonium fluoride,tbaf,tetrabutylazanium fluoride,tetrabutyl ammonium fluoride,tetra-n-butylammonium fluoride,tetrabutylamine, fluoride,n,n,n-tributylbutan-1-aminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride,n,n,n-tributyl-1-butanaminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride 1:1
2724141
CHEBI:51990
tetrabutylazanium;fluoride
CCCC[N+](CCCC)(CCCC)CCCC.[F-]
429-41-4,109-99-9,7732-18-5
C16H36FN
261.46
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylammonium fluoride,tbaf,tetrabutylazanium fluoride,tetrabutyl ammonium fluoride,tetra-n-butylammonium fluoride,tetrabutylamine, fluoride,n,n,n-tributylbutan-1-aminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride,n,n,n-tributyl-1-butanaminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride 1:1
2724141
CHEBI:51990
tetrabutylazanium;fluoride
CCCC[N+](CCCC)(CCCC)CCCC.[F-]
Linear Formula [CH3(CH2)3]4NF
Merck Index 15,9332
Formula Weight 261.46
Physical Form Solution
Concentration 0.90 to 1.10M
Density 0.8870g/mL
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.887
Water 6% max.
Color Brown to Green
Quantity 100mL
Flash Point −17°C
Chemical Name or Material Tetrabutylammonium fluoride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
May cause drowsiness or dizzines

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 207-057-2