Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium fluoride, 1M solution in THF, AcroSeal™

Catalog No. p-4228980
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 429-41-4 | C16H36FN | 261.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium fluoride
Color Brown to Green
CAS 429-41-4, 109-99-9, 7732-18-5
Physical Form Solution
Quantity 100 mL
Molecular Formula C16H36FN
Linear Formula [CH3(CH2)3]4NF
MDL Number MFCD00011747
Flash Point −17°C
Merck Index 15,9332
Synonym tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
InChI Key FPGGTKZVZWFYPV-UHFFFAOYSA-M
SMILES [F-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium;fluoride
Molecular Weight (g/mol) 261.47
PubChem CID 2724141
ChEBI CHEBI:51990
Formula Weight 261.46
Concentration or Composition (by Analyte or Components) 0.90 to 1.10M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.887
Density 0.8870g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.