Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium fluoride, 75% solution in water, Acros Organics

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
429-41-4
C16H36FN
261.46
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylammonium fluoride,tbaf,tetrabutylazanium fluoride,tetrabutyl ammonium fluoride,tetra-n-butylammonium fluoride,tetrabutylamine, fluoride,n,n,n-tributylbutan-1-aminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride,n,n,n-tributyl-1-butanaminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride 1:1
2724141
CHEBI:51990
tetrabutylazanium;fluoride
CCCC[N+](CCCC)(CCCC)CCCC.[F-]
429-41-4
C16H36FN
261.46
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylammonium fluoride,tbaf,tetrabutylazanium fluoride,tetrabutyl ammonium fluoride,tetra-n-butylammonium fluoride,tetrabutylamine, fluoride,n,n,n-tributylbutan-1-aminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride,n,n,n-tributyl-1-butanaminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride 1:1
2724141
CHEBI:51990
tetrabutylazanium;fluoride
CCCC[N+](CCCC)(CCCC)CCCC.[F-]
Specific Gravity 0.953
Density 0.9530g/mL
Quantity 25g
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NF
Merck Index 15,9332
Concentration or Composition (by Analyte or Components) 75% in aqueous solution
Formula Weight 261.46
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium fluoride

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS Signal Word: Danger

EINECSNumber : 207-057-2