Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium fluoride, 75% solution in water

Catalog No. p-4813676
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 429-41-4 | C16H36FN | 261.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429-41-4
Molecular Formula C16H36FN
Molecular Weight (g/mol) 261.47
MDL Number MFCD00011747
InChI Key FPGGTKZVZWFYPV-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
PubChem CID 2724141
ChEBI CHEBI:51990
SMILES [F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 0.9530g/mL
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NF
Quantity 25 g
Merck Index 15,9332
Specific Gravity 0.953
IUPAC Name tetrabutylazanium;fluoride
Formula Weight 261.46
Concentration or Composition (by Analyte or Components) 75% in aqueous solution
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium fluoride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
EINECSNumber 207-057-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.