Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Hydroxide, 0.20 Normal (N/5), Reagents

Catalog No. 01333468
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-468 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-468 Supplier Reagents Holdings Llc Supplier No. CT0040001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabutylammonium Hydroxide, 0.20 Normal (N/5), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 67-63-0 , 67-56-1 , 2052-49-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.01
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Hydroxide, 0.20 Normal (N/5)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.