Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Hydroxide, 1.0 Molar in Methanol, Reagents

Catalog No. 01333467
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-467 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-467 Supplier Reagents Holdings Llc Supplier No. CT005600100C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabutylammonium Hydroxide, 1.0 Molar in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 2052-49-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless to Slight Yellow
Boiling Point 65°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 0.83
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Hydroxide, 1.0 Molar in Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.