Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Hydroxide (10% in Methanol) [for non-aqueous titration], TCI America™
SDP

Catalog No. T004125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T004125ML 25 mL
T0041500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T004125ML Supplier TCI America Supplier No. T004125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
IUPAC Name tetrabutylazanium hydroxide
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Color Colorless
Quantity 25 mL
UN Number 2924
Formula Weight 259.48
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Hydroxide (10% in Methanol) [for non-aqueous titration]
EINECSNumber (2)-0186
RTECSNumber BS5425000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.