Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium hydroxide, 1M solution in methanol, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
2.5L
Packaging:
Plastic bottle
2052-49-5,67-56-1
C16H37NO
259.478
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
2052-49-5,67-56-1
C16H37NO
259.478
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
Phase transfer catalyst
Linear Formula [CH3(CH2)3]4NOH
Density 0.9100g/mL
Formula Weight 259.46
Packaging Plastic bottle
Refractive Index 1.3775
Solubility Solubility in water: soluble.
Specific Gravity 0.91
Boiling Point 65.0°C
Melting Point -98.0°C
Quantity 100mL
Flash Point 12°C
Chemical Name or Material Tetrabutylammonium hydroxide, 1M Solutionin Methanol
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 218-147-6

RTECSNumber : BS5425000

TSCA : TSCA