Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water

Catalog No. p-3737443
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC176610500 50 g Plastic bottle
AC176612500 250 g Plastic bottle
AC176610010 1 kg Plastic bottle
AC176610025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC176610500 Supplier Thermo Scientific Chemicals Supplier No. 176610500
Only null left
Add to Cart

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium hydroxide
Name Note 40 wt.% Solution in Water
CAS 2052-49-5, 7732-18-5
Color White to Yellow
Density 0.9950g/mL
Boiling Point >100.0°C
Packaging Plastic bottle
Molecular Formula C16H37NO
Linear Formula [CH3(CH2)3]4NOH
Quantity 50 g
MDL Number MFCD00009425
Beilstein 04, II, 634
Fieser 05,645; 11,500
Specific Gravity 0.995
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
Solubility Information Solubility in water: soluble.
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium;hydroxide
Molecular Weight (g/mol) 259.48
PubChem CID 2723671
Formula Weight 259.46
Percent Purity 38 to 42% (Total base)
Physical Form Crystals or Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.