Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium hydroxide, 40 wt.% solution in methanol, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
2052-49-5,67-56-1
C16H37NO
259.478
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
2052-49-5,67-56-1
C16H37NO
259.478
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
Phase transfer catalyst
Flash Point 12°C
Packaging Glass bottle
Quantity 100mL
Boiling Point 65.0°C
Color Colorless to Yellow
Melting Point -98.0°C
Density 0.9100g/mL
Specific Gravity 0.91
Linear Formula [CH3(CH2)3]4NOH
Beilstein 04, II, 634
Fieser 05,645; 11,500
Formula Weight 259.46
Physical Form Solution
Percent Purity ≥40%
Chemical Name or Material Tetrabutylammonium hydroxide, 40 wt.% solution in methanol
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger