Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium iodide, 98%

Catalog No. p-4510114
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle

CAS: 311-28-4 | C16H36IN | 369.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 311-28-4
Molecular Formula C16H36IN
Molecular Weight (g/mol) 369.38
MDL Number MFCD00011636
InChI Key DPKBAXPHAYBPRL-UHFFFAOYSA-M
Synonym tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
PubChem CID 67553
IUPAC Name tetrabutylazanium;iodide
SMILES [I-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 141°C to 150°C
Color White to Cream
Ignition Residue 1% max.
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NI
Quantity 25 g
Fieser 05,646; 06,566; 07,355; 09,447; 11,501
Solubility Information Solubility in water: <5%
Formula Weight 369.36
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium iodide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 206-220-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.