Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium phosphate monobasic, 0.4M solution in acetonitrile, AcroSeal™

Catalog No. ac435991000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435991000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC435991000 Supplier Thermo Scientific Chemicals Supplier No. 435991000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium phosphate monobasic
CAS 5574-97-0, 75-05-8
Melting Point -48.0°C
Color Colorless to Yellow
Density 0.8280g/mL
Boiling Point 81.0°C to 82.0°C
Flash Point 2°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C16H38NO4P
Linear Formula (CH3(CH2)3)4NH2PO4
MDL Number MFCD00064526
Quantity 100 mL
Specific Gravity 0.828
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
Solubility Information Solubility in water: soluble.
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name dihydrogen phosphate;tetrabutylazanium
Molecular Weight (g/mol) 339.46
PubChem CID 2735142
Viscosity 0.35 Pas (20°C)
Formula Weight 339.45
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.