Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracaine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607321
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
20 kg
Packaging:
Amber Glass Bottle
Poly Pail

TE220, 94-24-6, C15H24N2O2

Spectrum™ Chemical Tetracaine, USP is a local anesthetic used to prevent pain and induce anesthesia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 94-24-6
Molecular Formula C15H24N2O2
Molecular Weight (g/mol) 264.37
InChI Key GKCBAIGFKIBETG-UHFFFAOYSA-N
IUPAC Name 2-(dimethylamino)ethyl 4-(butylamino)benzoate
SMILES CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.