Learn More
Tetrachlorophthalic anhydride, 98%
Description
Protection of amines as their tetrachlorophthalimide derivatives, by reaction with the anhydride in the presence of triethylamine, has been found to be useful in oligosaccharide chemistry. The group is stable to a wide range of reagents but can be selectively cleaved with ethylenediamine at lower temperatures than for the parent phthalimide derivatives; in particular, carboxylic esters are not cleaved under these conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Protection of amines as their tetrachlorophthalimide derivatives, by reaction with the anhydride in the presence of triethylamine, has been found to be useful in oligosaccharide chemistry. The group is stable to a wide range of reagents but can be selectively cleaved with ethylenediamine at lower temperatures than for the parent phthalimide derivatives; in particular, carboxylic esters are not cleaved under these conditions.
Solubility
Hydrolyzes slowly in water.
Notes
Hygroscopic. Store away from oxidizing agents, acids.
Specifications
Specifications
| Melting Point | 254°C to 258°C |
| Boiling Point | 349°C to 354°C |
| Flash Point | 362°C (683°F) |
| Odor | Weak |
| Quantity | 2500 g |
| UN Number | UN3077 |
| Beilstein | 211560 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Hydrolyzes slowly in water. |
| Formula Weight | 285.9 |
| Show More |
Safety and Handling
| Hazard Category | H317-H318-H334-H350 |
| Hazard Statement | GHS H Statement H334-H318-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P305+P351+P338-P308+P313-P310-P333+P313-P342+P311-P363-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 204-171-4 |
| RTECSNumber | TI3450000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.