Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroterephthalonitrile 98.0+%, TCI America™
SDP

Catalog No. T1058100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1058100MG 100 mg
T10585G 5 g
T105825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1058100MG Supplier TCI America Supplier No. T1058100MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1897-41-2
Molecular Formula C8Cl4N2
Molecular Weight (g/mol) 265.902
MDL Number MFCD00059583
InChI Key TXRVDQMSXQKAPG-UHFFFAOYSA-N
Synonym Perchloroterephthalonitrile
PubChem CID 74694
IUPAC Name 2,3,5,6-tetrachlorobenzene-1,4-dicarbonitrile
SMILES C(#N)C1=C(C(=C(C(=C1Cl)Cl)C#N)Cl)Cl

Specifications

Quantity 100 mg
Physical Form Crystal-Powder at 20°C
UN Number 3077
Formula Weight 265.90
Percent Purity ≥98.0% (GC)
Chemical Name or Material Tetrachloroterephthalonitrile
EINECSNumber (3)-4603
RTECSNumber TI8275000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.