Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetracosane 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Specifications
| Melting Point | 51°C |
| Color | White |
| Boiling Point | 250°C |
| Quantity | 100 g |
| Formula Weight | 338.66 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Tetracosane |
Safety and Handling
| EINECSNumber | (2)-0010 |
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction