Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracosanoic acid, 99%

Catalog No. AC215640050
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

CAS: 557-59-5 | C24H48O2 | 368.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-59-5
Molecular Formula C24H48O2
Molecular Weight (g/mol) 368.63
MDL Number MFCD00002810
InChI Key QZZGJDVWLFXDLK-UHFFFAOYSA-N
Synonym lignoceric acid, unii-rk3vcw5y1l, n-tetracosanoic acid, rk3vcw5y1l, n-tetracosanoate, lignozerinsaeure, tetracosansaeure, carnaubic acid, tetracosoic acid, tetracosanic acid
PubChem CID 11197
ChEBI CHEBI:28866
IUPAC Name tetracosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 84.0°C to 86.0°C
Color White
Boiling Point 272.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)22CO2H
Quantity 5 g
Beilstein 02, I, 181
Merck Index 15, 5541
Solubility Information Solubility in water: .. Other solubilities: practically insoluble in ethanol, soluble in hot hydrocarbons and hot ketones
Formula Weight 368.63
Percent Purity 99%
Physical Form Shiny Crystalline Powder or Flakes
Chemical Name or Material Tetracosanoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
EINECSNumber 209-180-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.