Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracyclo[6.2.1.1(3,6).0(2,7)]dodec-4-ene 96.0+%, TCI America™
SDP

Catalog No. T181725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T181725G 25 g
T18171G 1 g
T18175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T181725G Supplier TCI America Supplier No. T181725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21635-90-5
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00213460
InChI Key XBFJAVXCNXDMBH-UHFFFAOYSA-N
Synonym 1,2,3,4,4a,5,8,8a-Octahydro-1,4:5,8-dimethanonaphthalene
PubChem CID 136844
SMILES C1CC2CC1C3C2C4CC3C=C4

Specifications

Color Colorless
Boiling Point 93°C
Quantity 25 g
Formula Weight 160.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetracyclo[6.2.1.1(3,6).0(2,7)]dodec-4-ene
EINECSNumber (4)-1683
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.