Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracyline hydrochloride, MP Biomedicals™

Catalog No. ICN10301125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10301125 25 g
ICN10301105 5 g
ICN10301180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10301125 Supplier MP Biomedicals, Inc Supplier No. 0210301125
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-75-5
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.898
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
Synonym tetracycline hydrochloride
PubChem CID 129628373
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 480.9
Physical Form Crystalline Powder
Chemical Name or Material Tetracyline hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.