Alfa Aesar™ Tetradecanoic acid, 98%

Manufacturer: Alfa Aesar™ A1206736

Quantity 500g

 View more versions of this product

Catalog No. AAA1206736

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 544-63-8
    Molecular Formula CH3(CH2)12CO2H
    Formula Weight 228.38
    MDL Number MFCD00002744
    Beilstein 508624
    Merck Index 14,6333
    Synonym Myristic acid
    Chemical Name or Material Tetradecanoic acid

    Specifications

    Formula Weight 228.38
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Myristic acid
    Health Hazard 1 Warning
    Flash Point 175°C (347°F)
    MDL Number MFCD00002744
    Merck Index 14,6333
    Molecular Formula CH3(CH2)12CO2H
    CAS 544-63-8
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 53° to 56°C
    RTECS Number QH4375000
    TSCA TSCA
    Beilstein 508624
    Boiling Point 326°C
    Quantity 500g
    Chemical Name or Material Tetradecanoic acid
    Assay 98%

    Description

    Applications
    It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.

    Solubility
    Insoluble in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit