Learn More
Tetradecanoic acid, 98%
Description
It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications
Specifications
| Melting Point | 53°C to 56°C |
| Density | 0.862 |
| Boiling Point | 326°C |
| Flash Point | 175°C (347°F) |
| Quantity | 2500 g |
| Beilstein | 508624 |
| Merck Index | 14,6333 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 228.38 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H319 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 208-875-2 |
| RTECSNumber | QH4375000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.