Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecanoic acid, 98%

Catalog No. p-7081868
Click to view available options
Quantity:
250 g
500 g
2500 g

CAS: 544-63-8 | C14H28O2 | 228.38 g/mol

It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-63-8
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00002744
InChI Key TUNFSRHWOTWDNC-UHFFFAOYSA-N
Synonym myristic acid, n-tetradecanoic acid, n-tetradecoic acid, crodacid, n-tetradecan-1-oic acid, 1-tridecanecarboxylic acid, hydrofol acid 1495, univol u 316s, emery 655, myristinsaeure
PubChem CID 11005
ChEBI CHEBI:28875
IUPAC Name tetradecanoic acid
SMILES CCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 53°C to 56°C
Density 0.862
Boiling Point 326°C
Flash Point 175°C (347°F)
Quantity 250 g
Beilstein 508624
Merck Index 14,6333
Solubility Information Insoluble in water.
Formula Weight 228.38
Percent Purity 98%
Chemical Name or Material Tetradecanoic acid
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 208-875-2
RTECSNumber QH4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.