Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecanophenone 98.0+%, TCI America™
SDP

Catalog No. T080425G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4497-05-6
Molecular Formula C20H32O
Molecular Weight (g/mol) 288.48
MDL Number MFCD00008985
InChI Key LXUIUVLDNRQBQJ-UHFFFAOYSA-N
Synonym Myristophenone, Phenyl Tridecyl Ketone
PubChem CID 78248
IUPAC Name 1-phenyltetradecan-1-one
SMILES CCCCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 206°C
Quantity 25 g
Formula Weight 288.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetradecanophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.