Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethoxysilane, 98%

Catalog No. p-7022271
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 78-10-4 | C8H20O4Si | 208.329 g/mol

Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.

Solubility
Soluble in ethanol and 2-propanol.

Notes
Degrades in water. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-10-4
Molecular Formula C8H20O4Si
Molecular Weight (g/mol) 208.329
MDL Number MFCD00009062
InChI Key BOTDANWDWHJENH-UHFFFAOYSA-N
Synonym tetraethyl orthosilicate, tetraethoxysilane, teos, ethyl silicate, silicon ethoxide, silicon tetraethoxide, silane, tetraethoxy, dynasil a, tetraethoxysilicon, ethyl orthosilicate
PubChem CID 6517
IUPAC Name tetraethyl silicate
SMILES CCO[Si](OCC)(OCC)OCC

Specifications

Melting Point -85°C
Density 0.934
Boiling Point 168°C
Flash Point 46°C (116°F)
Odor Pleasant
Assay Percent Range 98%
Refractive Index 1.383
Linear Formula (C2H5O)4Si
Quantity 100 mL
UN Number UN1292
Beilstein 1422225
Sensitivity Moisture sensitive
Merck Index 14,3851
Solubility Information Soluble in ethanol and 2-propanol.
Formula Weight 208.33
Percent Purity 98%
Chemical Name or Material Tetraethoxysilane
Show More Show Less
Hazard Category H226-H319-H332-H335
Hazard Statement GHS H Statement
H226-H332-H319-H335
Flammable liquid and vapour.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAETHYL SILICATE
EINECSNumber 201-083-8
RTECSNumber VV9450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.