Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium Bromide (Certified), Fisher Chemical

Catalog No. o4602500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 71-91-0
Molecular Formula C8H20BrN
Molecular Weight (g/mol) 210.159
MDL Number MFCD00011825
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
PubChem CID 6285
IUPAC Name tetraethylazanium;bromide
SMILES CC[N+](CC)(CC)CC.[Br-]

Specifications

Color White
Density 1.4g/cm³
pH 6.5
Assay Percent Range ≥98 %
Packaging Glass Bottle
Linear Formula (C2H5)4NBr
Quantity 500 g
Formula Weight 210.16
Percent Purity ≥98%
Grade Certified
Physical Form Solid
Chemical Name or Material Tetraethylammonium Bromide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.