Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium hexafluorophosphate, 98%

Catalog No. p-7049615
Click to view available options
Quantity:
10 g
50 g

CAS: 429-07-2 | C8H20F6NP | 275.219 g/mol

It is used extensively in electrolytes for electrochemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429-07-2
Molecular Formula C8H20F6NP
Molecular Weight (g/mol) 275.219
MDL Number MFCD00043179
InChI Key KLKUOIXSIDDDCN-UHFFFAOYSA-N
Synonym tetraethylammonium hexafluorophosphate, ammonium, tetraethyl-, hexafluorophosphate 1-, et4npf6, n,n,n-triethylethanaminium hexafluorophosphate, tetraethylammoniumhexafluorophosphate, tetraethylazanium hexafluorophosphate, tetraethylammonium hexafluorophosphate v, ethanaminium,n,n-triethyl-, hexafluorophosphate 1-, ethanaminium, n,n,n-triethyl-, hexafluorophosphate 1-
PubChem CID 9885
IUPAC Name tetraethylazanium;hexafluorophosphate
SMILES CC[N+](CC)(CC)CC.F[P-](F)(F)(F)(F)F

Specifications

Quantity 10 g
UN Number UN2811
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,and acetonitrile.
Formula Weight 275.23
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Tetraethylammonium hexafluorophosphate
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 207-056-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.