Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetraethylammonium hydroxide, 35% w/w aq. soln.
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless to Yellow |
Density | 1.023 g/mL |
Refractive Index | 1.404 |
Linear Formula | (CH3CH2)4NOH |
Quantity | 500 g |
UN Number | UN3267 |
Merck Index | 14,9201 |
Solubility Information | Miscible with water. |
Formula Weight | 147.26 |
Concentration or Composition (by Analyte or Components) | 35% w/w aq. soln. |
Show More |
Safety and Handling
Hazard Category | H290-H300-H311-H314-H335-H370-H372 |
Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
Precautionary Statement | P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S. |
EINECSNumber | 201-073-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'
missing translation for 'spotOppurtunityImprovement'missing translation for 'shareContentCorrection'