Alfa Aesar™ Tetraethylammonium perchlorate, 98% (dry wt.), cont. »10% water

Manufacturer: Alfa Aesar™ 4406209

Quantity 10g

 View more versions of this product

Catalog No. AA4406209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2567-83-1
    Molecular Formula (C2H5)4NClO4
    Formula Weight 229.71
    MDL Number MFCD00041977
    UN Number UN1479
    Beilstein 3638093
    Chemical Name or Material Tetraethylammonium perchlorate

    Specifications

    Physical Form Wet Crystals
    Formula Weight 229.71
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula (C2H5)4NClO4
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    MDL Number MFCD00041977
    CAS 2567-83-1
    Beilstein 3638093
    Concentration Contains ≈ 10% water
    Quantity 10g
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    UN Number UN1479
    Melting Point >300°C
    Odor Odorless
    Chemical Name or Material Tetraethylammonium perchlorate
    Assay 98%, (dry wt.)

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit