Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylene glycol, 99.5%

Catalog No. AC149590025
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum

CAS: 112-60-7 | C8H18O5 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-60-7
Molecular Formula C8H18O5
Molecular Weight (g/mol) 194.23
InChI Key UWHCKJMYHZGTIT-UHFFFAOYSA-N
Synonym tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843
PubChem CID 8200
ChEBI CHEBI:44920
IUPAC Name 2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCO)O

Specifications

Melting Point -4.1°C
Color Yellow
Density 1.1200g/mL
Boiling Point 324.0°C to 330.0°C
Flash Point 176°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4580 to 1.4600
Linear Formula O(CH2CH2OCH2CH2OH)2
Quantity 2.5 L
Beilstein 01,468
Specific Gravity 1.12
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in benzene and toluene, practically insoluble in diethylether
Viscosity 44.9 mPa.s (25°C)
Formula Weight 194.23
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Tetraethylene glycol
Show More Show Less
EINECSNumber 203-989-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.