Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylene glycol, 99%

Catalog No. AAB238800F
Click to view available options
Quantity:
500 mL
2500 mL

CAS: 112-60-7 | C8H18O5 | 194.227 g/mol

Tetraethylene glycol is used as a solvent to dissolve water-insoluble compounds. It is employed as a nitrocellulose solvent, a plasticizer, a natural gas desiccant, a hydraulic fluid and in lacquers and coatings. Furthermore, it is used in the synthesis of tetraethylene glycol methacrylate monomer. It acts as a chemical intermediate and acts as a solvent for the manufacturing of inks and dyes. In addition, it serves as a coupling agent in the production of textile lubricants and formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylene glycol is used as a solvent to dissolve water-insoluble compounds. It is employed as a nitrocellulose solvent, a plasticizer, a natural gas desiccant, a hydraulic fluid and in lacquers and coatings. Furthermore, it is used in the synthesis of tetraethylene glycol methacrylate monomer. It acts as a chemical intermediate and acts as a solvent for the manufacturing of inks and dyes. In addition, it serves as a coupling agent in the production of textile lubricants and formulations.

Solubility
Miscible with water, ethanol, acetone, acetic acid, glycerin, pyridine, aldehydes, carbon tetrachloride and diethyl ether. Immiscible with oil, benzene, toluene, fat and aliphatic hydrocarbons.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-60-7
Molecular Formula C8H18O5
Molecular Weight (g/mol) 194.227
MDL Number MFCD00002879
InChI Key UWHCKJMYHZGTIT-UHFFFAOYSA-N
Synonym tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843
PubChem CID 8200
ChEBI CHEBI:44920
IUPAC Name 2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCO)O

Specifications

Melting Point -4°C
Density 1.125
pH 8.5
Boiling Point 313°C to 314°C
Flash Point 193°C (379°F)
Odor Odorless
Refractive Index 1.459
Quantity 2500 mL
Beilstein 1634320
Solubility Information Miscible with water,ethanol,acetone,acetic acid,glycerin,pyridine,aldehydes,carbon tetrachloride and diethyl ether. Immiscible with oil,benzene,toluene,fat and aliphatic hydrocarbons.
Formula Weight 194.23
Percent Purity 99%
Chemical Name or Material Tetraethylene glycol
Show More Show Less
Hazard Category H315-H320-H341-H360-H373
Precautionary Statement P201-P202-P262-P264b-P280-P302+P352-P305+P351+P338-P308+P313-P314-P332+P313-P362-P501c
EINECSNumber 203-989-9
RTECSNumber XC2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.