Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetraethylenepentamine, technical
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -40°C |
Density | 0.9900g/mL |
Boiling Point | 340°C |
Flash Point | 139°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Refractive Index | 1.5055 |
Linear Formula | HN(CH2CH2NHCH2CH2NH2)2 |
Quantity | 2.5 kg |
Specific Gravity | 0.99 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful if swallowed. May cause an allergic skin reaction. Toxic in contact with skin. Toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously wit |
EINECSNumber | 203-986-2 |
RTECSNumber | KH8585000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction