Learn More
Tetraethyltin, 98%
Description
Tetraethyltin is used in the electronics industry. Used as catalyst for olefin polymerization and to produce other organotin compounds. Also used as a preservative for wood, textiles, paper, and leather.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -112°C |
Density | 1.187 g/mL |
Boiling Point | 181°C |
Flash Point | 53°C (128°F) |
Odor | Characteristic |
Refractive Index | 1.473 |
Quantity | 10 g |
UN Number | UN3384 |
Solubility Information | Insoluble in water; Soluble in alcohol and ether |
Formula Weight | 234.94 |
Show More |
Safety and Handling
Hazard Category | H226-H300+H310+H330 |
Hazard Statement | GHS H Statement H300-H310-H330-H226 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S. |
EINECSNumber | 209-906-2 |
RTECSNumber | WH8625000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.