Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluorophthalic Acid 98.0+%, TCI America™
SDP

Catalog No. T098625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T098625G 25 g
T09865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T098625G Supplier TCI America Supplier No. T098625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 652-03-9
Molecular Formula C8F4O4
Molecular Weight (g/mol) 236.08
MDL Number MFCD00002407
InChI Key YJLVXRPNNDKMMO-UHFFFAOYSA-L
Synonym tetrafluorophthalic acid, 1,2-benzenedicarboxylic acid, 3,4,5,6-tetrafluoro, 3,4,5,6-tetrafluorophthalic, 2,3,4,5-tetrafluorophthalic acid, 3,4,5,6-tetrafluorobenzene-1,2-dicarboxylic acid, 2,3,4,5-tetrafluoro-phthalic acid, 3,4,5,6-tetrafluoro-phthalic acid, tetrafluorobenzene-1,2-dicarboxylic acid, 3,4,5,6-tetrafluoro-1,2-benzenedicarboxylic acid, pubchem1407
PubChem CID 69544
IUPAC Name tetrafluorobenzene-1,2-dicarboxylate
SMILES [O-]C(=O)C1=C(F)C(F)=C(F)C(F)=C1C([O-])=O

Specifications

Melting Point 154°C
Quantity 25 g
Formula Weight 238.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluorophthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.