Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahexylammonium Iodide 98.0+%, TCI America™
SDP

Catalog No. T101025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T101025G 25 g
T10105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T101025G Supplier TCI America Supplier No. T101025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2138-24-1
Molecular Formula C24H52IN
Molecular Weight (g/mol) 481.591
MDL Number MFCD00041981
InChI Key VRKHAMWCGMJAMI-UHFFFAOYSA-M
Synonym tetrahexylammonium iodide, tetra-n-hexylammonium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide, tetrahexylazanium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide 1:1, acmc-209fjk, tetrahexyl ammonium iodide, tetra-n-hexyl ammonium iodide, ammonium, tetrahexyl-, iodide, 1-hexanaminium,n,n-trihexyl-, iodide
PubChem CID 75056
IUPAC Name tetrahexylazanium;iodide
SMILES CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[I-]

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 25 g
Formula Weight 481.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrahexylammonium Iodide
EINECSNumber (2)-0186
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.