Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydro-4H-pyran-4-one, 99%

Catalog No. p-4509654
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 29943-42-8 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29943-42-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
InChI Key JMJRYTGVHCAYCT-UHFFFAOYSA-N
Synonym tetrahydro-4h-pyran-4-one, 4-oxotetrahydropyran, tetrahydropyran-4-one, dihydro-2h-pyran-4 3h-one, 4h-pyran-4-one, tetrahydro, 4-thp, tetrahydro-2h-pyran-4-one, tetrahydro-pyran-4-one, 4-oxotetrahydopyran, tetrahydro-.gamma.-pyrone
PubChem CID 121599
IUPAC Name oxan-4-one
SMILES C1COCCC1=O

Specifications

Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 166.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530
Quantity 5 g
Beilstein 17,I,131
Specific Gravity 1.08
Solubility Information Solubility in water: miscible
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetrahydro-4H-pyran-4-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 249-967-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.