Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrahydrocannabinol Antibody (THC3), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP307897DL594
Description
Tetrahydrocannabinol Monoclonal antibody specifically detects Tetrahydrocannabinol in Non-species specific samples. It is validated for ELISASpecifications
Tetrahydrocannabinol | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Non-species specific | |
Purified |
ELISA | |
THC3 | |
ELISA | |
C21H30O2;Delta9-Tetrahydrocannabinol;Delta9-THC;Deltanyne;Dronabinol;Marinol;Tetrahydrocannabinol | |
Tetrahydrocannabinol (THC) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction