Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, 99.8%, Acros Organics

Click to view available options
Quantity:
1L
2.5L
Packaging:
AcroSeal™ Glass bottle
109-99-9,128-37-0
C4H8O
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
109-99-9,128-37-0
C4H8O
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran,furan, tetrahydro,butylene oxide,furanidine,hydrofuran,tetramethylene oxide,oxacyclopentane,1,4-epoxybutane,tetrahydrofuranne,tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
Assay Percent Range 99.8%
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Formula Weight 72.11
Percent Purity 99.8%
Grade Biochemical
Density 0.8800g/mL
Packaging AcroSeal™ Glass bottle
Refractive Index 1.407
Solubility Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Specific Gravity 0.88
Flash Point −21°C
Viscosity 0.55 mPa.s (20°C)
Boiling Point 66.0°C
Melting Point -108.4°C
Quantity 1L
Chemical Name or Material Tetrahydrofuran
Show More Show Less