Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™

Catalog No. p-7077656
Click to view available options
Quantity:
100 mL
1 L
2.5 L

CAS: 109-99-9 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9, 128-37-0
Molecular Formula C4H8O
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108.4°C
Density 0.8800g/mL
Boiling Point 66.0°C
Quantity 100 mL
Flash Point −21°C
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.407
Specific Gravity 0.88
Viscosity 0.55 mPa.s (20°C)
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Molecular Weight (g/mol) 72.107
Formula Weight 72.11
Percent Purity 99.85%
Grade Extra Dry
Chemical Name or Material Tetrahydrofuran
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H statement
May cause respiratory irritation
Causes serious eye irritation
Highly flammable liquid and vapor
May form explosive peroxides
Suspected of causing cancer
Precautionary Statement GHS P statement

Keep away from heat/sparks/open flames/hot surfaces
No smoking
Wear protective gloves/protective clothing/eye protection/face protection
Avoid breathing dust/fume/gas/mist/vapors/spray
If in eyes; rinse cautiously with water for several minutes
Remove contact lenses, if present and easy to do. Continue rinsing.  Wash with plenty of soap and water
Keep container tightly closed
EINECSNumber 203-726-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.