Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT

Catalog No. AC181500250
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

Tetrahydrofuran, >-99.85%, C4H8O, CAS Number-109-99-9, 128-37-0, tetraidrofurano, furanidine, tetrahydrofuranne, butylene oxide, tetrahydrofuran, furan, tetrahydro, oxacyclopentane, hydrofuran, 1,4-epoxybutane, tetramethylene oxide, 1L, 99.9%, 17, 10, CHEBI:26911, Undesignated, 0.8800g/mL | CAS: 109-99-9 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9, 128-37-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Density 0.8800g/mL
Quantity 25 L
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.4060 to 1.4080 (20°C, 589nm)
Assay Percent Range 99.9%
Beilstein 17, 10
Water 0.006% max. (Coulometric)
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Merck Index 15, 9356
Formula Weight 72.11
Percent Purity ≥99.85%
Grade Extra Pure
Chemical Name or Material Tetrahydrofuran, Anhydrous, Stabilized
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Precautionary Statement GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.