Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, 99.9%, for HPLC, unstabilized, Thermo Scientific Chemicals

Catalog No. ac610100040
Click to view available options
Quantity:
4 L
Packaging:
EcoSafPak™

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9, 128-37-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Absorbance 1 max. at 210nm, 0.6 max. at 215nm, 0.3 max. at 230nm, 0.1 max. at 254nm, (1 cm cell vs water)
Density 0.8800g/mL
Quantity 4 L
Identification Pass Test
Packaging EcoSafPak™
Refractive Index 1.4040 - 1.4070
Residue after Evaporation 5ppm max.
Assay Percent Range 99.9%
Vapor Pressure Vapor Pressure: 200mbar at 20°C
Beilstein 17, 10
Water 0.02% max.
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Merck Index 15, 9356
Formula Weight 72.11
Percent Purity ≥99.9%
Grade HPLC
Chemical Name or Material Tetrahydrofuran, 99.9%
Show More Show Less
Hazard Category Danger

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.