Tetrahydrofuran, anhydrous, >99.8%%, Alfa Aesar™

For Synthesis and Liquid Chromatography

Manufacturer: Alfa Aesar 44608AE

Quantity 100mL

 View more versions of this product

Catalog No. AA44608AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-99-9
    Physical Form Liquid
    Assay Percent Range >99.8%
    Molecular Formula C4H8O
    Formula Weight 72.11
    MDL Number MFCD00005356
    Chemical Name or Material Tetrahydrofuran, Water <100ppm

    Specifications

    Formula Weight 72.11
    Quantity 100mL
    Color Colorless
    DOT Information DOT Class: 3, Packaging Group: II
    EINECS Number 203-726-8
    Physical Form Liquid
    Odor Ether-like
    Vapor Pressure 200 hPa (150mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range >99.8%
    Molecular Formula C4H8O
    MDL Number MFCD00005356
    CAS 109-99-9
    Health Hazard 1 H225-H351-H319-H335: Highly flammable liquid and vapour. Suspected of causing cancer. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Tetrahydrofuran, Water <100ppm

    H225-H351-H319-H335: Highly flammable liquid and vapour. Suspected of causing cancer. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit