Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9%, Solstice

Catalog No. 6001687 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-016-87 19 L Metal Can
60-016-88 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-016-87 Supplier Solstice Supplier No. PP34119
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

CAS Max % 100.0000%
Density 0.8892 g/cm3
Quantity 19 L
Flash Point −14°C
Packaging Metal Can
Water 0.05% max.
UN Number UN2056
Beilstein 102391
Boiling Point 66°C
Solubility Information Miscible in Water at 20°C
Formula Weight 72.11
Percent Purity >99.9%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Tetrahydrofuran, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Tetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.